siRNA Details
Gene Symbol: Klk1b21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010642.2 | NP_034772.1 | 3 | 433 | ||
GenBank mRNA | AB039276.1 | - | 3 | 436 | ||
AK144221.1 | - | 3 | 400 | |||
BC012243.1 | - | 3 | 367 | 4167772 | ||
BC145383.1 | - | 3 | 389 | 9053941 |
