siRNA Details
Gene Symbol: Klk1b8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008457.2 | NP_032483.1 | 4 | 557 | ||
GenBank mRNA | BC031798.1 | - | 4 | 562 | 4167600 | |
X03994.1 | - | 4 | 556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
