siRNA Details
Gene Symbol: Klra12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010646.1 | NP_034776.1 | 3 | 263 | ||
GenBank mRNA | AF204265.1 | - | 3 | 215 | ||
AF204266.1 | - | 3 | 215 | |||
AF204267.1 | - | 3 | 206 | |||
AF204268.1 | - | 3 | 206 | |||
AF307947.1 | - | 3 | 267 | |||
AF307948.1 | - | 3 | 263 |
Gene Symbol: Klra23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024470.1 | NP_077790.1 | 4 | 338 | ||
GenBank mRNA | AF074459.1 | - | 4 | 338 | ||
AF074463.1 | - | 4 | 329 | |||
AF283250.1 | - | 3 | 221 | |||
AF283251.1 | - | 3 | 212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
