siRNA Details
Gene Symbol: Klra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008464.2 | NP_032490.1 | 5 | 529 | ||
GenBank mRNA | U10092.1 | - | 5 | 529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
