siRNA Details
Gene Symbol: Klra7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF074457.1 | - | 2, 3 | 216 | ||
AF283248.1 | - | 2, 3 | 216 | |||
AF283249.1 | - | 2, 3 | 216 | |||
AF288375.1 | - | 2, 3 | 209 | |||
AF288376.1 | - | 2, 3 | 238 | |||
AF307945.1 | - | 2, 3 | 216 | |||
AF307946.1 | - | 2, 3 | 216 | |||
AF419249.1 | - | 498 | ||||
BC099859.1 | - | 2, 3 | 215 | 40040027 | ||
BC099860.1 | - | 2, 3 | 215 | 40040024 | ||
BC099862.1 | - | 2, 3 | 215 | 40040021 | ||
BC130251.1 | - | 2, 3 | 264 | 40142464 | ||
BC140984.1 | - | 2, 3 | 262 | 9055596 | ||
GU434662.1 | - | 2, 3 | 209 | |||
GU434663.1 | - | 2, 3 | 209 | |||
S78689.1 | - | 2, 3 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
