siRNA Details
Gene Symbol: Klra7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110323.1 | NP_001103793.1 | 3, 4 | 334 | ||
NM_014194.5 | NP_055009.5 | 2, 3 | 274 | |||
GenBank mRNA | AF074457.1 | - | 2, 3 | 211 | ||
AF283248.1 | - | 2, 3 | 211 | |||
AF283249.1 | - | 2, 3 | 211 | |||
AF288375.1 | - | 2, 3 | 204 | |||
AF288376.1 | - | 2, 3 | 233 | |||
AF307945.1 | - | 2, 3 | 211 | |||
AF307946.1 | - | 2, 3 | 211 | |||
AF419249.1 | - | 493 | ||||
AK143240.1 | - | 2, 3 | 234 | |||
BC099859.1 | - | 2, 3 | 210 | 40040027 | ||
BC099860.1 | - | 2, 3 | 210 | 40040024 | ||
BC099862.1 | - | 2, 3 | 210 | 40040021 | ||
BC130251.1 | - | 2, 3 | 259 | 40142464 | ||
BC140984.1 | - | 2, 3 | 257 | 9055596 | ||
EU429321.1 | - | 2, 3 | 204 | |||
GU434662.1 | - | 2, 3 | 204 | |||
GU434663.1 | - | 2, 3 | 204 | |||
S78689.1 | - | 2, 3 | 233 | |||
U10093.1 | - | 3, 4 | 334 | |||
U10094.1 | - | 2, 3 | 274 | |||
U10095.1 | - | 3, 4 | 334 | |||
U12890.1 | - | 2, 3 | 225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
