siRNA Details
Gene Symbol: Klrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010652.2 | NP_034782.1 | 2, 3 | 354 | ||
GenBank mRNA | AF109784.1 | - | 2, 3 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
