siRNA Details
Gene Symbol: Klrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136068.2 | NP_001129540.1 | 7 | 1626 | ||
NM_001288664.1 | NP_001275593.1 | 6 | 1524 | |||
NM_010652.2 | NP_034782.1 | 6 | 1575 | |||
GenBank mRNA | AF106008.1 | - | 7 | 1589 | ||
AK138801.1 | - | 7 | 1583 |
Gene Symbol: Klrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036555.1 | - | 1433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
