siRNA Details
Gene Symbol: Kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102411.1 | NP_001095881.1 | 4 | 637 | ||
NM_001102412.1 | NP_001095882.1 | 4 | 637 | |||
NM_023125.3 | NP_075614.1 | 4 | 637 | |||
GenBank mRNA | AK005547.1 | - | 4 | 602 | ||
AY462058.1 | - | 4 | 517 | |||
AY660571.1 | - | 4 | 544 | |||
BC018158.1 | - | 4 | 589 | 4162960 | ||
BC108936.1 | - | 4 | 593 | 40039851 | ||
D84415.1 | - | 4 | 517 | |||
D84435.1 | - | 4 | 517 |
