siRNA Details
Gene Symbol: Krt33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013570.1 | NP_038598.1 | 1 | 304 | ||
GenBank mRNA | AA681979.1 | - | 1 | 304 | ||
AI466315.1 | - | 1 | 45 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
