siRNA Details
Gene Symbol: Krt27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010666.1 | NP_034796.1 | 5 | 905 | ||
GenBank mRNA | AB013607.1 | - | 5 | 905 | ||
AK077384.1 | - | 5 | 917 | |||
BC140996.1 | - | 5 | 929 | 9055608 | ||
BC140997.1 | - | 5 | 929 | 9055609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
