siRNA Details
Gene Symbol: Krt6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008476.3 | NP_032502.3 | 4 | 901 | ||
GenBank mRNA | AK009394.1 | - | 4 | 904 | ||
BC064470.1 | - | 4 | 875 | |||
BC080820.1 | - | 4 | 901 | 30141946 | ||
K02108.1 | - | 4 | 893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
