siRNA Details
Gene Symbol: Krtap12-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010670.1 | NP_034800.1 | 1 | 223 | ||
GenBank mRNA | BC107319.1 | - | 1 | 210 | 40053395 | |
BC107320.1 | - | 1 | 210 | 40053396 |
Gene Symbol: Gm10024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081452.2 | NP_001074921.1 | 1 | 165 |
Gene Symbol: Gm10100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205033.1 | NP_001191962.1 | 1 | 154 |
Gene Symbol: Gm10142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205035.1 | NP_001191964.1 | 1 | 154 |
Gene Symbol: Gm10272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA079946.1 | - | 1 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
