siRNA Details
Gene Symbol: Krtap13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010671.1 | NP_034801.1 | 1 | 516 | ||
GenBank mRNA | AF031485.1 | - | 1 | 516 | ||
AK003960.1 | - | 1 | 539 | |||
BC063754.1 | - | 1 | 523 | 30293948 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
