siRNA Details
Gene Symbol: Krtap19-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010676.2 | NP_034806.2 | 1 | 124 | ||
GenBank mRNA | AA717139.1 | - | 1 | 132 | ||
D86422.1 | - | 1 | 135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
