siRNA Details
Gene Symbol: Ky | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024291.3 | NP_077253.3 | 4, 5 | 351 | ||
GenBank mRNA | AK047076.1 | - | 4, 5 | 742 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108180, XM_006243657.
Gene Ontology Categories:
