siRNA Details
Gene Symbol: Lad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133664.3 | NP_598425.2 | 3 | 1051 | ||
GenBank mRNA | AK083526.1 | - | 3 | 1052 | ||
BC016257.1 | - | 3 | 722 | 4912735 | ||
BC025580.1 | - | 3 | 955 | 5325039 | ||
BC031131.1 | - | 3 | 898 | 4954891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
