siRNA Details
Gene Symbol: Dsg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181564.2 | NP_853543.1 | 11 | 1677 | ||
GenBank mRNA | AB240192.1 | - | 10 | 1860 | ||
AY170623.1 | - | 4 | 625 | |||
AY191584.1 | - | 11 | 1677 | |||
AY227349.1 | - | 11 | 1543 | |||
BC109009.2 | - | 11 | 1677 | |||
BC150475.2 | - | 11 | 1677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
