siRNA Details
Gene Symbol: Lcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008490.2 | NP_032516.2 | 5 | 659 | ||
GenBank mRNA | AI574112.1 | - | 3 | 399 | ||
AK004914.1 | - | 5 | 655 | |||
AK149476.1 | - | 5 | 656 | |||
BC028861.1 | - | 5 | 642 | 4212194 | ||
J05154.1 | - | 5 | 639 |
