siRNA Details
Gene Symbol: Ldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113408.1 | NP_001106879.1 | 4 | 560 | ||
NM_010697.1 | NP_034827.1 | 4 | 745 | |||
GenBank mRNA | AB221569.1 | - | 3 | 102 | ||
AB250383.1 | - | 3 | 99 | |||
AF030333.1 | - | 4 | 263 | |||
BC013624.1 | - | 4 | 385 | 3708169 | ||
CK790183.1 | - | 4 | 565 | |||
U69270.1 | - | 4 | 745 | |||
U70375.1 | - | 3 | 95 | |||
U89488.1 | - | 4 | 343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
