siRNA Details
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145953.1 | NP_001139425.1 | 4, 5 | 602 | ||
NM_010705.3 | NP_034835.1 | 4, 5 | 575 | |||
GenBank mRNA | AK008593.1 | - | 3, 4 | 244 | ||
AK089249.1 | - | 4, 5 | 519 | |||
AK149783.1 | - | 4, 5 | 549 | |||
AK155244.1 | - | 5, 6 | 421 | |||
AK155484.1 | - | 4, 5 | 517 | |||
AK159536.1 | - | 4, 5 | 543 | |||
AK170803.1 | - | 4, 5 | 632 | |||
BC132328.1 | - | 4, 5 | 528 | 40130605 | ||
BC138790.1 | - | 4, 5 | 528 | 8861812 | ||
BC145419.1 | - | 4, 5 | 528 | 9053977 | ||
J03723.1 | - | 2, 3 | 475 | |||
M33215.1 | - | 2, 3 | 184 | |||
X16074.1 | - | 4, 5 | 622 | |||
X16834.1 | - | 4, 5 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
