siRNA Details
Gene Symbol: Lhx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290646.1 | NP_001277575.1 | 5 | 1373 | ||
NM_010710.4 | NP_034840.1 | 5 | 1434 | |||
GenBank mRNA | AF124734.1 | - | 5 | 1159 | ||
AK053572.1 | - | 5 | 1435 | |||
BC055741.1 | - | 5 | 1158 | 6413339 | ||
BC086638.1 | - | 5 | 1373 |

More Information
Other Species Targeted

Gene Ontology Categories:
