siRNA Details
Gene Symbol: Lhx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008499.5 | NP_032525.1 | 2 | 675 | ||
GenBank mRNA | AK033927.1 | - | 2 | 662 | ||
AK048778.1 | - | 2 | 664 | |||
AK140208.1 | - | 2 | 676 | |||
BC057585.1 | - | 2 | 563 | 6414270 | ||
U61155.1 | - | 2 | 791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
