siRNA Details
Gene Symbol: Lhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025565.2 | NP_001020736.1 | 4 | 1325 | ||
NM_001042577.1 | NP_001036042.1 | 4 | 1325 | |||
NM_010714.3 | NP_034844.1 | 6 | 952 | |||
GenBank mRNA | AB221560.1 | - | 5 | 835 | ||
AF113518.1 | - | 4 | 805 | |||
AF134761.1 | - | 6 | 951 | |||
AJ243851.1 | - | 6 | 951 | |||
AK012930.1 | - | 4 | 763 | |||
AK013209.1 | - | 5 | 977 | |||
AK031478.1 | - | 4 | 1327 | |||
AK086557.1 | - | 1 | 884 | |||
BC072623.1 | - | 4 | 1338 | 6825043 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
