siRNA Details
Gene Symbol: Lifr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113386.1 | NP_001106857.1 | 6, 7 | 961 | ||
NM_013584.2 | NP_038612.1 | 5, 6 | 610 | |||
GenBank mRNA | AK145864.1 | - | 6, 7 | 962 | ||
AK155524.1 | - | 5, 6 | 837 | |||
BC031929.1 | - | 5, 6 | 603 | 4159053 | ||
D17444.1 | - | 5, 6 | 617 | |||
D26177.1 | - | 5, 6 | 617 | |||
S73495.1 | - | 5, 6 | 617 | |||
S73496.1 | - | 5, 6 | 617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
