siRNA Details
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083188.1 | NP_001076657.1 | 22 | 2355 | ||
NM_001199310.1 | NP_001186239.1 | 22 | 2363 | |||
NM_010715.2 | NP_034845.2 | 22 | 2280 | |||
GenBank mRNA | AK153993.1 | - | 22 | 2242 | ||
AK160208.1 | - | 7 | 649 | |||
AK161793.1 | - | 15 | 1438 | |||
AK168643.1 | - | 22 | 2359 | |||
BC028287.1 | - | 22 | 2205 | 4911261 | ||
BC138541.1 | - | 22 | 2363 | 8861561 | ||
BC138542.1 | - | 22 | 2363 | 8861562 | ||
U04674.1 | - | 21 | 2148 | |||
U19604.1 | - | 22 | 2393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
