siRNA Details
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141813.1 | - | 13, 14 | 1748 | ||
AK170166.1 | - | 13, 14 | 2074 | |||
AK172083.1 | - | 12, 13 | 1507 | |||
BC063768.1 | - | 13, 14 | 1728 | |||
BQ828670.1 | - | 1, 2 | 115 | |||
U15159.1 | - | 13, 14 | 1743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
