siRNA Details
Gene Symbol: Lmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142335.1 | NP_001135807.1 | 4 | 586 | ||
NM_001142336.1 | NP_001135808.1 | 4 | 672 | |||
NM_001142337.1 | NP_001135809.1 | 4 | 596 | |||
NM_008505.4 | NP_032531.2 | 3 | 713 | |||
GenBank mRNA | AK013416.1 | - | 4 | 744 | ||
BC057880.1 | - | 3 | 777 | 5349568 | ||
CX226855.1 | - | 4 | 655 | |||
M64360.1 | - | 3 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
