siRNA Details
Gene Symbol: Phyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010726.2 | NP_034856.1 | 4 | 372 | ||
GenBank mRNA | AF023463.1 | - | 4 | 337 | ||
AK146753.1 | - | 4 | 357 | |||
AK151372.1 | - | 4 | 320 | |||
AK164495.1 | - | 4 | 323 | |||
AK165412.1 | - | 4 | 316 | |||
AK168648.1 | - | 4 | 342 | |||
AK169213.1 | - | 4 | 314 | |||
BC002018.1 | - | 4 | 349 | 3489939 | ||
D88670.1 | - | 4 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
