siRNA Details
Gene Symbol: Sh2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028796.1 | - | 6 | 1289 | ||
AK156499.1 | - | 5 | 1168 | |||
AK157297.1 | - | 5 | 1158 | |||
AK165124.1 | - | 4 | 397 | |||
AK171128.1 | - | 6 | 1232 | |||
AK172551.1 | - | 5 | 1158 | |||
AK172573.1 | - | 5 | 1162 | |||
BC006759.1 | - | 5 | 1142 | 3709861 | ||
U89992.2 | - | 1053 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
