siRNA Details
Gene Symbol: Lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286181.1 | NP_001273110.1 | 7 | 1274 | ||
NM_001286182.1 | NP_001273111.1 | 7 | 1231 | |||
NM_010728.3 | NP_034858.2 | 6 | 1651 | |||
GenBank mRNA | AK135213.1 | - | 6 | 782 | ||
AK141995.1 | - | 6 | 1537 | |||
AK159199.1 | - | 6 | 1454 | |||
AK159241.1 | - | 6 | 1430 | |||
AK159263.1 | - | 6 | 1491 | |||
AK159368.1 | - | 6 | 1413 | |||
AK159796.1 | - | 6 | 1425 | |||
AK160096.1 | - | 6 | 1425 | |||
AK164634.1 | - | 7 | 1232 | |||
AK166279.1 | - | 6 | 1359 | |||
AK166386.1 | - | 1495 | ||||
AK166442.1 | - | 1651 | ||||
BC018439.1 | - | 6 | 1502 | 2655752 | ||
M65142.1 | - | 6 | 1419 | |||
M65143.1 | - | 3 | 278 |
