siRNA Details
Gene Symbol: Xcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008510.1 | NP_032536.1 | 3 | 215 | ||
GenBank mRNA | AK079820.1 | - | 3 | 228 | ||
BC062249.1 | - | 3 | 215 | 3974601 | ||
D43769.1 | - | 3 | 215 | |||
U15607.1 | - | 3 | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
