siRNA Details
Gene Symbol: Lrmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281980.1 | NP_001268909.1 | 3 | 292 | ||
NM_001281981.1 | NP_001268910.1 | 4 | 333 | |||
NM_008511.3 | NP_032537.3 | 3 | 564 | |||
GenBank mRNA | AK040180.1 | - | 3 | 565 | ||
AK050646.1 | - | 3 | 1002 | |||
AK079932.1 | - | 3 | 522 | |||
AK153823.1 | - | 3 | 562 | |||
AK153865.1 | - | 3 | 294 | |||
AK160009.1 | - | 3 | 524 | |||
AK169578.1 | - | 4 | 317 | |||
AY753324.1 | - | 2 | 127 | |||
BC052909.1 | - | 3 | 500 | 30076553 | ||
U10484.1 | - | 3 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
