siRNA Details
Gene Symbol: Lrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008514.4 | NP_032540.2 | 9 | 2248 | ||
GenBank mRNA | AF074265.1 | - | 9 | 2117 | ||
BC060704.1 | - | 9 | 2248 | 6409058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
