siRNA Details
Gene Symbol: Lrrfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008515.4 | NP_032541.1 | 17 | 1452 | ||
GenBank mRNA | AF045573.1 | - | 17 | 1389 | ||
BC144955.1 | - | 8 | 789 | |||
BC144956.1 | - | 19 | 1410 | 9053504 | ||
BC145642.1 | - | 19 | 1410 | 40130945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
