siRNA Details
Gene Symbol: Lrrfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111312.1 | NP_001104782.1 | 9 | 919 | ||
NM_008515.4 | NP_032541.1 | 19 | 1662 | |||
GenBank mRNA | AF045573.1 | - | 19 | 1599 | ||
AK041162.1 | - | 9 | 870 | |||
BC144955.1 | - | 10 | 999 | |||
BC144956.1 | - | 21 | 1620 | 9053504 | ||
BC145642.1 | - | 21 | 1620 | 40130945 |

More Information
Other Species Targeted

Gene Ontology Categories:
