siRNA Details
Gene Symbol: Lrrfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008515.4 | NP_032541.1 | 6, 7 | 537 | ||
GenBank mRNA | AF045573.1 | - | 6, 7 | 474 | ||
AK044174.1 | - | 6, 7 | 416 | |||
CJ154753.1 | - | 6, 7 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
