siRNA Details
Gene Symbol: Klrb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030599.4 | NP_085102.4 | 3 | 305 | ||
GenBank mRNA | AF324826.1 | - | 3 | 281 | ||
AF338321.1 | - | 3 | 236 | |||
AF338322.1 | - | 3 | 236 | |||
AF342896.1 | - | 3 | 284 | |||
AF354259.1 | - | 242 | ||||
AF354260.1 | - | 242 | ||||
AK030702.1 | - | 3 | 254 | |||
AK041228.1 | - | 3 | 306 | |||
AK138766.1 | - | 3 | 304 | |||
AK169820.1 | - | 3 | 322 | |||
BC132126.1 | - | 3 | 280 | 40130403 | ||
BC132128.1 | - | 3 | 280 | 40130405 | ||
BC145389.1 | - | 3 | 280 | 9053947 | ||
DQ237928.1 | - | 3 | 238 | |||
HQ713433.1 | - | 236 | ||||
M77677.1 | - | 3 | 281 | |||
X64719.1 | - | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
