siRNA Details
Gene Symbol: Ly6c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252055.1 | NP_001238984.1 | 3 | 241 | ||
NM_001252056.1 | NP_001238985.1 | 3 | 226 | |||
NM_001252057.1 | NP_001238986.1 | 3 | 226 | |||
NM_001252058.1 | NP_001238987.1 | 2 | 312 | |||
NM_010741.3 | NP_034871.2 | 3 | 226 | |||
GenBank mRNA | AI893484.1 | - | 3 | 193 | ||
AK088782.1 | - | 3 | 203 | |||
AK143577.1 | - | 3 | 218 | |||
BC010764.1 | - | 3 | 219 | 4221342 | ||
BE448813.1 | - | 2 | 312 | |||
BY118129.1 | - | 3 | 200 | |||
CB595639.1 | - | 3 | 229 | |||
CK031980.1 | - | 3 | 192 |
Gene Symbol: Ly6c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099217.1 | NP_001092687.1 | 2 | 183 | ||
GenBank mRNA | BC092082.1 | - | 2 | 183 | 4983733 | |
D86232.1 | - | 2 | 304 | |||
M18466.1 | - | 2 | 184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
