siRNA Details
Gene Symbol: Ly6e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164036.1 | NP_001157508.1 | 3 | 831 | ||
NM_001164037.1 | NP_001157509.1 | 4 | 704 | |||
NM_001164038.1 | NP_001157510.1 | 4 | 620 | |||
NM_001164039.1 | NP_001157511.1 | 4 | 617 | |||
NM_001164040.1 | NP_001157512.1 | 3 | 414 | |||
NM_008529.3 | NP_032555.1 | 3 | 828 | |||
GenBank mRNA | AK014207.1 | - | 3 | 286 | ||
BC002116.1 | - | 3 | 231 | 3490293 | ||
BC003926.1 | - | 3 | 234 | 3485382 | ||
BC005684.1 | - | 3 | 266 | 3155116 | ||
BC019113.1 | - | 3 | 257 | 2651183 | ||
BC037143.1 | - | 3 | 242 | |||
BC040691.1 | - | 3 | 251 | 5025135 | ||
BC053523.1 | - | 3 | 234 | 4205673 | ||
BC080711.1 | - | 3 | 231 | 3969173 | ||
CT010220.1 | - | 2 | 153 | |||
CT010244.1 | - | 2 | 153 | |||
U04268.1 | - | 2 | 196 | |||
U09192.1 | - | 3 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
