siRNA Details
Gene Symbol: Ly75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013825.3 | NP_038853.2 | 8 | 1327 | ||
GenBank mRNA | AF395445.1 | - | 8 | 1327 | ||
AK049301.1 | - | 8 | 1370 | |||
AK144922.1 | - | 8 | 1380 | |||
AK157921.1 | - | 8 | 1391 | |||
BC150734.1 | - | 1348 | 9087645 | |||
U19271.1 | - | 1361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
