siRNA Details
Gene Symbol: Tmed1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010744.3 | NP_034874.2 | 3 | 456 | ||
GenBank mRNA | AK133513.1 | - | 3 | 456 | ||
AK160525.1 | - | 2 | 359 | |||
AK166092.1 | - | 2 | 358 | |||
BC006715.1 | - | 2 | 350 | 3154568 | ||
BC016568.1 | - | 2 | 596 | |||
BC138498.1 | - | 3 | 504 | 8861518 | ||
BC138499.1 | - | 3 | 503 | 8861519 | ||
BC145298.1 | - | 3 | 469 | |||
U41805.1 | - | 3 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
