siRNA Details
Gene Symbol: Ly86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010745.2 | NP_034875.1 | 2 | 283 | ||
GenBank mRNA | AB007599.1 | - | 2 | 283 | ||
AK027929.1 | - | 2 | 272 | |||
BC065783.1 | - | 2 | 188 | |||
BC096414.1 | - | 2 | 251 | 849991 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
