siRNA Details
Gene Symbol: Ly96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159711.1 | NP_001153183.1 | 1, 2 | 163 | ||
NM_016923.2 | NP_058619.1 | 1, 2 | 163 | |||
GenBank mRNA | AB018550.1 | - | 1, 2 | 163 | ||
AF168120.1 | - | 1, 2 | 133 | |||
AK019283.1 | - | 1, 2 | 187 | |||
AY641431.1 | - | 1, 2 | 111 | |||
BC116785.1 | - | 1, 2 | 159 | 40126104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
