siRNA Details
Gene Symbol: Lyz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017372.3 | NP_059068.1 | 2 | 274 | ||
GenBank mRNA | AK148516.1 | - | 2 | 275 | ||
AK150998.1 | - | 2 | 277 | |||
AK153244.1 | - | 2 | 277 | |||
AK153475.1 | - | 2 | 275 | |||
AK159276.1 | - | 2 | 324 | |||
AK159640.1 | - | 2 | 271 | |||
BC002069.1 | - | 2 | 266 | 3593360 | ||
BC054463.1 | - | 2 | 266 | 6514977 |
Gene Symbol: Lyz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013590.4 | NP_038618.1 | 2 | 274 | ||
GenBank mRNA | BC061129.1 | - | 2 | 274 | 30296274 | |
X51547.1 | - | 2 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
