siRNA Details
Gene Symbol: Lyz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK150998.1 | - | 4 | 1219 | ||
AK153244.1 | - | 4 | 1219 | |||
AK159276.1 | - | 4 | 1266 | |||
AK159640.1 | - | 4 | 1213 | |||
BC002069.1 | - | 4 | 1208 | 3593360 | ||
BC019611.1 | - | 901 | ||||
BC054463.1 | - | 4 | 1208 | 6514977 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
