siRNA Details
Gene Symbol: Tm4sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008536.3 | NP_032562.1 | 5, 6 | 1023 | ||
GenBank mRNA | AK077326.1 | - | 3, 4 | 526 | ||
AK133556.1 | - | 524 | ||||
AK134603.1 | - | 4, 5 | 959 | |||
AK164718.1 | - | 3, 4 | 527 | |||
BC083073.1 | - | 5, 6 | 1023 | 6337376 | ||
KC508089.1 | - | 3, 4 | 498 | |||
L15429.1 | - | 3, 4 | 513 | |||
L15443.1 | - | 3, 4 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
