siRNA Details
Gene Symbol: Mxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010751.3 | NP_034881.2 | 2, 3 | 320 | ||
GenBank mRNA | AK151014.1 | - | 2, 3 | 282 | ||
AK151114.1 | - | 2, 3 | 281 | |||
AK151305.1 | - | 2, 3 | 319 | |||
AK151882.1 | - | 2, 3 | 281 | |||
AK153303.1 | - | 2, 3 | 281 | |||
BC034860.1 | - | 2, 3 | 257 | 1448816 | ||
BY106585.1 | - | 2, 3 | 324 | |||
CT010233.1 | - | 2, 3 | 169 | |||
L38926.1 | - | 2, 3 | 205 | |||
U20614.1 | - | 2, 3 | 274 | |||
X83106.1 | - | 2, 3 | 218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
