siRNA Details
Gene Symbol: Smad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016769.4 | NP_058049.3 | 3 | 801 | ||
GenBank mRNA | AB008192.1 | - | 3 | 510 | ||
AF016189.1 | - | 3 | 777 | |||
AK041743.1 | - | 3 | 322 | |||
AK048626.1 | - | 3 | 801 | |||
AK083158.1 | - | 3 | 409 | |||
BC022904.1 | - | 3 | 944 | |||
BC066850.1 | - | 3 | 487 | 30432720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
