siRNA Details
Gene Symbol: Smad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008540.2 | NP_032566.2 | 12 | 2596 | ||
GenBank mRNA | AK170255.1 | - | 1 | 75 | ||
AY493561.1 | - | 12 | 2118 | |||
BC046584.1 | - | 12 | 2596 | 6313280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
